B24412.03
B24412.06
B24412.14
B24412.03EA
44.5
EUR
InStock
B24412.03
B24412.06
B24412.14
3-Methoxyphenylboronsäure ≥97%
3-Methoxyphenylboronsäure
Purity: 97%
CAS No.: 10365-98-7
Molecular Formula: C7H9BO3
Formula Weight: 151.96
Melting Point: 158-162°
Beilstein Reference: 2831223