B24472.22
B24472.36
B24472.22EA
32.2
EUR
InStock
B24472.22
B24472.36
Isobutyrylchlorid ≥98%
Isobutyrylchlorid
Purity: 98%
CAS No.: 79-30-1
Molecular Formula: (CH3)2CHCOCl
Formula Weight: 106.55
Flash Point: 8°(46°F)
Refractive Index: 1.4070
Odour: Unpleasant
Melting Point: ca -90°
Boiling Point: 91-93°
Beilstein Reference: 605393
UN No.: 2395
MDL No.: MFCD00000717