B24656.03
B24656.06
B24656.14
B24656.03EA
26.2
EUR
InStock
B24656.03
B24656.06
B24656.14
4-Ethylphenylboronsäure ≥97%
4-Ethylphenylboronsäure
Purity: 97%
CAS No.: 63139-21-9
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: 150-154°
Beilstein Reference: 3030638