B24678.22
B24678.06
B24678.14
B24678.22EA
580
EUR
InStock
B24678.22
B24678.06
B24678.14
(R)-(+)-2-Pyrrolidinon-5-carbonsäure ≥98%
(R)-(+)-2-Pyrrolidinon-5-carbonsäure
Purity: 98+%
CAS No.: 4042-36-8
Molecular Formula: C5H7NO3
Formula Weight: 129.12
Melting Point: 158-160°
Beilstein Reference: 82133