B24741.06
B24741.14
B24741.06EA
75
EUR
InStock
B24741.06
B24741.14
4-(Dimethylamino)zimtaldehyd ≥98%
4-(Dimethylamino)zimtaldehyd
Purity: 98%
CAS No.: 6203-18-5
Molecular Formula: C11H13NO
Formula Weight: 175.23
Melting Point: 137-144°
Beilstein Reference: 972369