B24750.06
B24750.14
B24750.22
B24750.06EA
41.1
EUR
InStock
B24750.06
B24750.14
B24750.22
Methyl-1,2,4-triazol-3-carboxylat ≥98%
Methyl-1,2,4-triazol-3-carboxylat
Purity: 98%
CAS No.: 4928-88-5
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
MDL No.: MFCD00135989