B24787.14
B24787.22
B24787.36
B24787.14EA
24.6
EUR
InStock
B24787.14
B24787.22
B24787.36
2,5-Dichlorphenol ≥98%
2,5-Dichlorphenol
Purity: 98%
CAS No.: 583-78-8
Molecular Formula: C6H4Cl2O
Formula Weight: 163.00
Odour: Phenol-like
Melting Point: 55-58°
Boiling Point: 211°
Beilstein Reference: 1907692
UN No.: 2020
MDL No.: MFCD00002174