B24832.22
B24832.36
B24832.22EA
46.6
EUR
InStock
B24832.22
B24832.36
Didecyldimethylammoniumbromid ≥80%, wässriges Gel
Didecyldimethylammonium bromide
CAS No.: 2390-68-3
Molecular Formula: C22H48BrN
Formula Weight: 406.54
Refractive Index: 1.4570
MDL No.: MFCD00012194