B25173.03
B25173.06
B25173.03EA
61.2
EUR
InStock
B25173.03
B25173.06
5-Iod-1,3-dimethyluracil ≥99%
5-Iod-1,3-dimethyluracil
Purity: 99%
CAS No.: 40738-83-8
Molecular Formula: C6H7IN2O2
Formula Weight: 266.04
Melting Point: 225° dec.
MDL No.: MFCD00192228