B25175.03
B25175.06
B25175.14
B25175.03EA
35.2
EUR
InStock
B25175.03
B25175.06
B25175.14
2-Amino-5-iodpyridin 97%
2-Amino-5-iodpyridin
Purity: 97%
CAS No.: 20511-12-0
Molecular Formula: C5H5IN2
Formula Weight: 220.01
Melting Point: 126-130°
Beilstein Reference: 108738