B25222.03
B25222.06
B25222.14
B25222.03EA
36.5
EUR
InStock
B25222.03
B25222.06
B25222.14
4-(Chlormethyl)-1,2-diphenylethan ≥98%
4-(Chlormethyl)-1,2-diphenylethan
Purity: 98%
CAS No.: 80676-35-3
Molecular Formula: C15H15Cl
Formula Weight: 230.73
Melting Point: 38-42°
UN No.: 1759
MDL No.: MFCD02093918