B25253.03
B25253.06
B25253.14
B25253.03EA
18.7
EUR
InStock
B25253.03
B25253.06
B25253.14
2-Brom-4-methylpyridin 96%
2-Brom-4-methylpyridin
Purity: 96%
CAS No.: 4926-28-7
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 110°(230°F)
Refractive Index: 1.5630
Boiling Point: 87°/10mm
Beilstein Reference: 107331
MDL No.: MFCD00082590