B25331.04
B25331.09
B25331.04EA
39.8
EUR
InStock
B25331.04
B25331.09
2-Brom-5-nitroanisol ≥98%
2-Brom-5-nitroanisol
Purity: 98%
CAS No.: 77337-82-7
Molecular Formula: C7H6BrNO3
Formula Weight: 232.03
Melting Point: 102-107°
Beilstein Reference: 2520496