B25515.06
B25515.14
B25515.22
B25515.06EA
51
EUR
InStock
B25515.06
B25515.14
B25515.22
(±)-2-Phenyl-3-butin-2-ol ≥98%
(±)-2-Phenyl-3-butin-2-ol
Purity: 98%
CAS No.: 127-66-2
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point: 96°(205°F)
Melting Point: 49-53°
Boiling Point: 92-93°/10mm
Beilstein Reference: 1100096
MDL No.: MFCD00004454