B25732.03
B25732.06
B25732.03EA
46.4
EUR
InStock
B25732.03
B25732.06
1H-Pyrazol-3-carbonsäure ≥97%
1H-Pyrazole-3-carboxylic acid
Purity: 97%
CAS No.: 1621-91-6
Molecular Formula: C4H4N2O2
Formula Weight: 112.09
Melting Point: 211° dec.
MDL No.: MFCD00077436