Sterilisation indicator strips
Lieferant: BAG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
75171
BAGA75171EA 185 EUR InStock
BAGA75171
Sterilisation indicator strips
Sterilisationsindikatoren
These indicator strips turns green to black colour.
  • Visible conformation of item exposed to dry heat sterilisation process
  • Monitors time and temperature
Zertifizierungen: Meets class I according to ISO 11140-1.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge