Water purification systems, Barnstead™ LabTower™ TII
Lieferant: BARNSTEAD THERMOLYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
50132141
BARN50132141EA 17700 EUR InStock
BARN50132141
Water purification systems, Barnstead™ LabTower™ TII
Wasseraufbereitungssysteme Reinstwassersysteme
These water purification systems convert tap water into high purity water (Type II) and provide an integrated storage solution. Designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity.
  • Three system options are available with permeate flows of 20, 40 or 60 L/hr
  • Stand-alone design with integrated 100 L reservoir takes up zero bench space
  • 5 μm prefilter and activated carbon in the combi-filter cartridge protect the RO membrane from free chlorine and particles
  • Hardness stabilising cartridge protects the reverse osmosis stage from hard water
  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Irradiation with UV light (TII units with UV option only) eliminates any bacteria or germs that are present
  • RS232 interface with adjustable send-interval for safe data transfer of all measured data to a PC computer or log printer
Typical applications: Rinsing lab glassware, supplying general lab equipment, preparing and diluting buffers, reagents and media.
Two ways to dispense water:
- Users can dispense directly from the system via the dispensing valve for calibration solutions, reagent preparation, filling of containers, general rinsing purposes, etc.
- Users can sispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.
Lieferumfang: All systems include an RO membrane, a high purity water cartridge, 10" 5 μm prefilter with hardness stabiliser cartridge, pressure regulator, 0,2 µm filter, integrated 100 L reservoir and UV lamp where applicable.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge