Thermo Scientific™ Digital Mini Rotator
Lieferant: BARNSTEAD THERMOLYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88882008
BARN88882008EA 1340 EUR InStock
BARN88882008
Thermo Scientific™ Digital Mini Rotator
Schüttler Rotations-/Wippschüttler
Provides smooth, accurate and orbital mixing of various vessels in the laboratory.
  • Precise speed control: ±1 min⁻¹ (≤100 min⁻¹), ±1% (101 to 450 min⁻¹)
  • Orbit diameter: 10 mm
  • 2 timer modes: Continuous, Timer
  • Multi-platform choices for loading all sizes vessels in the laboratory
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge