Thermo Scientific™ Digital Vortex Mixer
Lieferant: BARNSTEAD THERMOLYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88882010
BARN88882010EA 461 EUR InStock
BARN88882010
Thermo Scientific™ Digital Vortex Mixer
Schüttler Vortex-Schüttler
Designed for accurate speed control for continuous mixing or pulsing of samples.
  • Digital display of speed and time
  • Continuous and Touch modes
  • Precise speed control: ±5% min⁻¹
  • Quick set up to Max rpm:
  • 3 seconds 1500 min⁻¹ (Touch)
  • 6 seconds 3000 min⁻¹ (Continuous)
  • Time set: up to 99 minutes
  • 2 Timer modes: Down-counting for repeat tests and Timer: Continuous, time set as 0
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge