Thermo Scientific™ Basic Vortex Mixer
Lieferant: BARNSTEAD THERMOLYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88882012
BARN88882012EA 366 EUR InStock
BARN88882012
Thermo Scientific™ Basic Vortex Mixer
Schüttler Vortex-Schüttler
The Thermo Scientific™ basic vortex mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.
  • Digital speed display
  • Continuous and Touch modes
  • Convenient speed adjustments by knob control
  • Stable for long-term operation
  • Multiple accessories for different vessels
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge