Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator
Lieferant: BARNSTEAD THERMOLYNE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88882016
BARN88882016EA 1860 EUR InStock
BARN88882016
Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator
Schüttler Rotations-/Wippschüttler
The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.
  • Digital display of speed and time
  • Adjustable rocking angle: 95 - 180°
  • Timer range: 0 - 9999 mins
  • Optional multiple size drums offering different capacity of tube rotation
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge