Particulate filter, P3 R
Lieferant: BARTELS RIEGER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
922404
BART922404EA 34.3 EUR InStock
BART922404
Particulate filter, P3 R
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
The high-performance particle filter for filtering or purifying air has a high-quality housing made from plastic.
  • Particle separation efficiency of >99,95%
  • Low inhalation resistance
  • Low weight and compact design
The particle filter is designed for use in areas contaminated by dusts and aerosols.
Zertifizierungen: EN 133, DIN EN 148-1, EN 143:200
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge