Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Filter cartridges

Lieferant: BARTELS RIEGER

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

924000
BART924000EA 41.1 EUR InStock
BART924000
Filter cartridges
Filter Filterkartuschen
The combination filter 39 ASt is designed for use in areas contaminated by dusts and solvent vapours.

  • Combined filters combine the protection of a gas and particle filter in one product
  • The design guarantees the filtration of organic gases and vapours (boiling point >65 °C) and a particle separation efficiency >94,95%

Cartridge filters can only be used in combination with a filter adapter on face pieces with thread connector connection.

Zertifizierungen: According to DIN EN 14387:2008, Class A1 P2 RD. CE certified.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge