Racks for cryogenic vials, TruCool®
Lieferant: BIOCISION
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BCS-222
BCISBCS-222EA 339 EUR InStock
BCISBCS-222
Racks for cryogenic vials, TruCool®
Gestelle Gestelle für Kryoröhrchen
These polypropylene cryogenic racks feature a locking mechanism that allow one handed opening for self-standing cryogenic vials.
  • Accommodates both round bottom and self-standing vial formats
  • Racks have A to J row and 1 to 5 column indexing for easy organization
  • Stackable when not in use
Multicolour pack contains one each of blue, green, orange, purple and pink racks.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge