Insulinspritzen, BD Micro-Fine™
Lieferant: BECTON DICKINSON MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
320926 320927 324892 324891 324826 324825 324827 324824
BDAM320926EA 155 EUR InStock
BDAM320926 BDAM320927 BDAM324892 BDAM324891 BDAM324826 BDAM324825 BDAM324827 BDAM324824
Insulinspritzen, BD Micro-Fine™
Spritzen Spritzen, medizinisch
Micro-Fine™ is a single-use syringe for diabetes. It offers a more comfortable and convenient way to inject insulin with three different volumes.
  • Made from surgical stainless steel
  • Precision and comfort
  • Optimum shape and sharpness of the needle tip and bevel
  • Electro polished needle shaft
The white and orange protective cap conforms with sterility and a minimal dead space reduces the risk of air bubbles and prevents waste of insulin.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge