Critical care syringes, BD Vacutainer®
Lieferant: BECTON DICKINSON MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
364356
BDAM364356EA 342 EUR InStock
BDAM364356
Critical care syringes, BD Vacutainer®
Spritzen Spritzen, medizinisch
Critical care syringes can be used to measure arterial blood gas, ionised calcium, ionised magnesium, as well as perform critical care tests.
  • Permits aspiration
Syringes contain calcium balanced lithium heparin spray dried on the sides of the syringe.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge