Blood collection tubes, BD Microtainer®
Lieferant: BECTON DICKINSON MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
365975 365954 365993 365986 365988 365968 365964 365966
613-3969EA 452 EUR InStock
613-3969 BDAM365954 BDAM365993 BDAM365986 BDAM365988 BDAM365968 BDAM365964 BDAM365966
Blood collection tubes, BD Microtainer®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
These tubes are used for the collection of capillary blood from skin puncture.
  • Tubes come with a variety of additives for various tests
  • Provided with visible fill lines and are all colour-coded for ease identification
  • Tubes are also available for light sensitive analytes
BD Microtainer® tubes with BD Microgard™ closure have an integrated collection scoop, along with a wider diameter (8 mm) for ease of collection and improved mixing ensuring a quality sample is collected.
Additionally closure helps protect the healthcare worker from exposure to potentially infected blood with the 'twist assist' cap which reduces the risk of splatter upon removal of the cap.
It can also be used with a tube extender to increase the length to 75 mm, for use in 13×75 mm racks. BD Microtainer® tubes are also available with Flotop collector and closure plug.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge