DNA fingerprinting kit
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-104
BE-104EA 366 EUR InStock
BE-104
DNA fingerprinting kit
Lehrmittel Forensik-Lehrmittel Lehrmittel zur Fingerabdruckanalyse
This DNA fingerprinting kit allows students to carry out their own criminal investigation by comparing DNA samples collected from suspects to DNA collected at a pseudo-crime scene.
  • Introduces DNA fingerprinting technique
  • Teaches DNA fragmentation by specific enzymes
  • Introduction to agarose electrophoresis
Students will use specialised molecules to cut the DNA into fragments, which can then be visualised by a process known as electrophoresis. The resulting pattern, or fingerprint, can be compared and the guilty suspect identified.
Lieferumfang: Supplied with components needed for hands-on experimentation for six groups of 4 - 5 students or 24 - 30 students with teacher’s guide and separate student’s guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge