Experimentation kits, DNA ligation
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-308
BE-308EA 305 EUR InStock
BE-308
Experimentation kits, DNA ligation
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
Following restriction enzyme digestion of DNA molecules, researchers need to rejoin the ends of the DNA to generate recombinant DNA, a process known as ligation. Ligation of DNA is achieved with the bacterial enzyme T4 DNA ligase, which catalyses the formation of phosphodiester bonds. The DNA Ligation kit teaches students about ligation as they ligate several DNA fragments together to make larger pieces of DNA that are easily identified by agarose electrophoresis.
  • Teaches joining of DNA ends by a process known as ligation
Also required: Agarose electrophoresis equipment.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge