Experimentation kits, expression of a recombinant protein
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-312
BE-312EA 361 EUR InStock
BE-312
Experimentation kits, expression of a recombinant protein
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
The final goal in molecular biology is often the expression of a recombinant protein. The transformed plasmids can be used as templates by the bacteria to produce protein. Students learn about essential promoters and other elements necessary for successful protein expression in bacteria, including the differences between inducible and constitutive (unregulated) expression.The Expression of a Recombinant Protein kit allows students to express a protein either constitutively or under the control of an inducible promoter, which is activated with IPTG (isopropyl-beta-D-thiogalactopyranoside).
  • Teaches recombinant protein expression
  • Students learn about bacterial growth phases and inducible vectors
Also required: Shaking incubator, low speed centrifuge for 1,5 to 2 ml tubes, protein electrophoresis equipment, water bath or beaker and thermometer.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4-5 students or 24-30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge