Experimentation kits, enzyme analysis
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-405
BE-405EA 342 EUR InStock
BE-405
Experimentation kits, enzyme analysis
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
This lab activity involves analysis of an enzyme reaction using a specific enzyme substrate and inhibitor. Students study how the rate of enzyme reaction is dependent on substrate concentration and the influence of agents (inhibitors) and conditions that affects the enzyme reaction such as pH and temperature.
  • Enzyme kinetic and regulation of enzyme activity
  • Role of substrate concentration
  • Effects of pH and temperature on the reaction rate
  • Inhibition of enzyme reaction
Also required: Spectrophotometer and cuvettes or microplate reader and microplate.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge