Experimentation kits, protein fingerprinting
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-408
BE-408EA 342 EUR InStock
BE-408
Experimentation kits, protein fingerprinting
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
In this lab activity, students learn to perform simple protein isolation procedures to isolate the protein fingerprint from various fresh tissues. They will compare the protein fingerprints of 4 different tissues to understand that the function of a particular organ is due to the proteins that are localised to the specific organ. Also included in this kit are four dried protein samples (from mouse liver, brain, heart and lungs) to compare as a control, if fresh tissues cannot be obtained.
  • Manipulation of protein samples
  • Protein molecules as the building blocks of life
  • Protein electrophoresis for problem solving tasks
  • Protein diversity across the animal kingdom
Also required: Protein electrophoresis equipment.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge