Experimentation kits, conservation of genetic information
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-409
BE-409EA 329 EUR InStock
BE-409
Experimentation kits, conservation of genetic information
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
In this lab activity, students learn to perform carefully controlled experiments to generate protein fragments using a proteolysis enzyme and then analyse the protein fragments by electrophoresis. By analysis of protein fragmentation patterns, i.e. protein fingerprints, students learn about protein sequence, structure, and their conservation. Students resolve a set of 3 functionally identical protein samples selected from throughout the animal kingdom; including human, bovine and sheep. After generating fingerprints by electrophoresis, students examine the protein fingerprint of each sample to determine the degree of conservation.
  • Manipulation of protein samples
  • The concept of protein sequence
  • Time course dependent experiments
  • Use of protein electrophoresis for problem solving tasks
Also required: Protein electrophoresis equipment, incubator, water bath or beaker and thermometer.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge