Experimentation kits, size exclusion chromatography
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BE-414
BE-414EA 303 EUR InStock
BE-414
Experimentation kits, size exclusion chromatography
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
The Size Exclusion Chromatography kit teaches gel filtration or size exclusion chromatography and the use of this method in purification of biological samples. This method is based on separation of protein molecules based on their molecular size. This lab activity involves running size exclusion chromatography for separation of molecules based on their molecular sizes.
  • Principles of gel filtration or size exclusion chromatography
  • Factors affecting gel filtration chromatography
  • Hands-on chromatography lab activity
  • Role of gel filtration and strategies in protein purification
Also required: Clamp and stand.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge