Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bench top cell sorter, CytoFLEX SRT

Lieferant: BECKMAN COULTER

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

C71883 C71884 C71885 C71886 C71887 C71888 C71889 C71900 C71901 C71902 C71903 C71904 C71905 C71906 C85213 C85214
BECMC71883EA 0 EUR InStock
BECMC71883 BECMC71884 BECMC71885 BECMC71886 BECMC71887 BECMC71888 BECMC71889 BECMC71900 BECMC71901 BECMC71902 BECMC71903 BECMC71904 BECMC71905 BECMC71906 BECMC85213 BECMC85214
Bench top cell sorter, CytoFLEX SRT
Zellzähler und Durchflusszytometer Durchflusszytometer
If you need an approachable instrument for sorting multiple populations to generate high quality material for downstream assays, the CytoFLEX SRT benchtop cell sorter is quick to learn and easy to operate. This extension of the CytoFLEX platform is built on the same optical features as the flow cytometer, but equally important, it continues to adhere to the principles that made multicolor applications accessible to many biomedical research, core research, pharma and biotechs.

  • Exquisite sensitivity for multicolor applications, up to 15 fluorescent colors to define populations
  • Parameter matched to the CytoFLEX S V-B-Y-R series
  • Intuitive software to facilitate multicolor analysis, CytExpert for CytoFLEX SRT
  • Uses technology to increase ease of use by automating sort stream setup and maintenance throughout the sort
  • 4-way sorting, mixed mode sorting, ability to catch aborts and preserve precious cells
  • High recovery and post sort viability
  • Accurate single-cell deposition using the Cyclone Movement System, including index sorting
  • Sort into tubes, slides, or microplates

CytoFLEX SRT cell sorter is a benchtop sorter. It is capable of meeting requirements for a wide range of sorting needs. And like the CytoFLEX Platform, it includes innovative technologies that simplify the setup and operation, empowering investigators to focus on the research questions.

The Violet-Blue-Yellow Green-Red (V-B-Y-R) series has 15 fluorescent detectors when fully activated. The fully activated instrument includes five fluorescent channels from the 405 nm (Violet) laser, two from the 488 nm (Blue) laser, five from the 561 nm (Yellow Green) laser, and three from the 638 nm (Red) laser. It can be purchased with as few as five with an option to activate additional lasers and detectors with an activation key.

It is capable of complex sort logic with different combinations of sort settings on each of four streams, including the ability to catch aborts of one of the other streams. The CytoFLEX SRT cell sorter is designed for reliability and supported by teams of experts, remote support is ready using BeckmanConnect for on demand access to our technical specialists, perform many routine maintenance procedures independently.

Additional specifications:
Nozzle diameter: 100 µm
Signal processing: Digital system with 7-decade display
Drop dive frequency: Automatically set, manually adjustable between 30000 - 35000 Hz
Sorting performance: With a target population of 5% and a threshold rate of <10,000 events per second, >99% purity and >80% of Poisson's expected yield
Sorting modes: 4-way sorting; enrichment or purity or single, ability to catch aborts
Collection vessels: 12×75 mm tubes, 15 ml conical tubes, 96-well plates, 96-deep-well plates, 384-well plates, slides
Optical filter: 450/45, 525/40 (2), 585/42, 610/20 (2), 660/10 (2), 675/30, 690/50, 710/50, 712/25, 780/60 (3)

Zertifizierungen: CE, RUO. EN12469, BS EN12469, SNS 12469, NF-095, YY-0569, JIS K 3800, AS 1807.1:2009.

Bestellinformation: CytoFLEX SRT is supplied with CyExpert for SRT, computer, monitor and keyboard. An optional Biosafety Cabinet including aerosol evacuation or a standalone Aerosol Evacuation Unit is available.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge