Compact system for 500 ml extraction KEX 500 / KEX 500 F, behrotest®
Lieferant: BEHR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B00217739
BEHRB00217739EA 0 EUR InStock
BEHRB00217739
Compact system for 500 ml extraction KEX 500 / KEX 500 F, behrotest®
Anaerobe Systeme
behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction.
With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
  • Complete with reaction vessels, extraction spacers and coolers
  • The heating mantle in use is continuously variable and has an integrated magnetic stirrer.
  • Practical stand for holding condensers securely between extractions
  • Holders for secure fixation of extraction adapters after removal of the sample vessels
Extractors with specially developed lifting tube ('Böckerhoff' type) guarantee homogeneous results at all sample locations.
F models: The use of extractors with a stopcock does away with the need for any additional distillation after extraction.
W×D×H (in cm): 23×33×80
Controller: No
Power consumption (in W): 350
Current consumption (in A): 3
Sample positions: 1
Capacity (in ml): 1000
Flask size (in ml): 1000
Extractor size (in ml): 500
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge