H-B DURAC® Plus Liquid-in-Glass Laboratory Thermometer
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B60700-1500
BELAB60700-1500EA 0 EUR InStock
BELAB60700-1500
H-B DURAC® Plus Liquid-in-Glass Laboratory Thermometer
Thermometer Analoge Thermometer
Durable, partial and total immersion thermometers. Mercury free, these thermometers feature blue organic liquid fill against lead-free white back glass.
  • Traceable to NIST, individually serialised
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, dairies, breweries and distilleries
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge