Liquid-in-glass thermometer, angled, H-B DURAC®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B608102900
BELAB608102900EA 88.4 EUR InStock
BELAB608102900
Liquid-in-glass thermometer, angled, H-B DURAC®, SP Bel-Art
Thermometer Analoge Thermometer
Designed for use in all heating blocks, this total immersion thermometer features a 90° angle and an arrow at 37 °C.
  • Red organic liquid against white back glass
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge