Maximum registering, autoclave bi-metal thermometer, H-B DURAC®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B613150000
BELAB613150000EA 130 EUR InStock
BELAB613150000
Maximum registering, autoclave bi-metal thermometer, H-B DURAC®, SP Bel-Art
Thermometer Analoge Thermometer
This durable bi-metal thermometer has a maximum registering pointer (red) that is moved up by the temperature measuring pointer (black) to indicate the highest temperature achieved, and remains there until manually reset.
  • Threaded connection (¼" NPT) screws directly into in-line processes, can be used to mount thermometer on included stand
  • Waterproof and pressurized, also has a removable protective silicone sleeve to prevent moisture from making contact with the dial
  • Constructed from durable 304 stainless steel with a glass lens
  • Withstands pressure, high temperatures and rust
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Zertifizierungen: Traceable to NIST.
Lieferumfang: Supplied with multi-language instructions (English, French, German, Italian, Portuguese and Spanish).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge