Trigger sprayers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F116200050
BELAF116200050EA 98 EUR InStock
BELAF116200050
Trigger sprayers, SP Bel-Art
Verschlüsse Spritzverschlüsse
PP, durable, chemical resistant.
  • Stainless steel, spring-action pump in the trigger sprayer will not corrode with repeated exposure to 10% bleach disinfecting solution
  • Sprayer without adapter fits most bottles with a 28 mm opening
  • Adapter plus trigger sprayer adds approx. 140 mm height to a wash bottle
Trigger sprayers with adapters fit any Scienceware® bottle with an easy to fill, 53 mm wide closure and offer convenient, adjustable spray dispensing of a wide range of chemicals including bleach, ethanol, machine oil and water. Simple to install sprayer is perfect for GHS Labelled safety-vented wash bottles (series 12416), as well as OSHA compliant 16 and 32 oz. Scienceware® non-vented, wide-mouth wash bottles
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge