Wash bottles, wide neck, with coloured caps, labelled, Safety-Vented™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F116420624 F116420622 F116420623 F116420638 F116430238
BELAF116420624EA 83.5 EUR InStock
BELAF116420624 BELAF116420622 BELAF116420623 BELAF116420638 BELAF116430238
Wash bottles, wide neck, with coloured caps, labelled, Safety-Vented™, SP Bel-Art
Flaschen Spritzflaschen
LDPE, with PP spray cap
  • Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labeled wash bottles
  • Allows dispensing with bottle upright or inverted
  • Volatiles are vented to eliminate solvent drips
  • Pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment
Labeled with six of the most commonly used solvent names or label as per the need. Solvents compatible with LDPE can be used. Custom labeling is also available.
Zertifizierungen: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge