Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Labelling tapes, rainbow pack, Write-On™, SP Bel-Art

Lieferant: Bel-Art Products, a Part of SP

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

F134630600
BELAF134630600EA 272 EUR InStock
BELAF134630600
Labelling tapes, rainbow pack, Write-On™, SP Bel-Art
Klebebänder
These labelling tapes are made of paper with rubber adhesive and are assembled in rainbow pack.

  • Resistant to moisture and most solvents
  • Withstands a maximum temperature of +247 °C for 30 minutes; can be used down to −73 °C
  • Withstands a steam autoclaving cycle at +121 °C

Adheres to any clean surface, fluoropolymer resin covered materials without leaving a sticky residue. Pencil, ball-point pen or solvent ink marker remain legible under the most demanding laboratory conditions.

Length: 36,4 m

Order Now
 

PRODUKT

TECHNISCHE DATEN

EGÄNZENDE PRODUKTE

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge