Bottle brush, Cleanware™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F170680000
BELAF170680000EA 150 EUR InStock
BELAF170680000
Bottle brush, Cleanware™, SP Bel-Art
Bürsten
Uniquely designed, swiveling pivot head enables thorough cleaning of bottles and flasks including difficult curves and corners.
  • Natural bristles set in wood will not scratch glass
  • Swivel head fits through a 29/42 standard taper neck, or any non-tapered neck with a diameter of 24 mm (1") or larger
  • Brush head is slightly convex, 45 mm (L) × 21 mm (W) with bristles 8 mm long
  • Brush head is attached to hardwood handle 37 cm long × 7 mm diameter
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge