Microcentrifuge tube floating racks, Prep-Safe™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F187420001
BELAF187420001EA 54 EUR InStock
BELAF187420001
Microcentrifuge tube floating racks, Prep-Safe™, SP Bel-Art
Gestelle Reagenzglasgestelle
PP
Unique tabs protect the samples from loss and contamination. Place tubes in slots and slide under tabs to keep tubes in place and caps closed when under pressure, floating or submerged. Use in water baths, ice or on the benchtop. Holds twenty eight 1,5 or 2,0 ml microcentrifuge tubes in numbered locations. Raised handle allows rack to be lifted without agitating samples.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge