Cryo storage box 0 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F188462000
BELAF188462000EA 644 EUR InStock
BELAF188462000
Cryo storage box 0 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Polycarbonate body and white polycarbonate lid, gel-filled.
  • Adapters change well size to hold 0,5 ml microcentrifuge tubes
  • Small footprint is ideal for benchtop use
  • Epoxy coated wire handle fastens lid and allows easy transport
  • All polycarbonate cooler body and lid clean easily
A non-toxic freezing gel is isolated within the walls of the cooler and lid, ensuring lasting low temperatures and protecting any labels or marks on the vials. The lid is indexed and marked to show tube location that matches locations printed on the cooler. Coolers can be stacked to save space and maximize freezer volume and have rubber feet to avoid slipping. Ideal for safeguarding samples in freezers with defrost cycle and fluctuating temperatures. Keep thirty-two 1,5 ml microcentrifuge tubes below 0 °C for 3 hours on the benchtop.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge