Freezer storage box, 100 places, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F188510016
BELAF188510016EA 136 EUR InStock
BELAF188510016
Freezer storage box, 100 places, SP Bel-Art
Boxen Kryoboxen
Freezer storage box provide compact, safe storage for tubes and vials.
  • Polypropylene construction
  • Available in a variety of colour choices
  • Hinged lids with snap tabs
  • Alphanumeric labeling
Great for organisation on the bench, in the freezer, or during transport, they can be used at temperatures –80 to +121 °C (112 to +249 °F)
Hinged lids with snap tab open easily and close securely, while clear translucent colour always lets you see inside. Opaque black boxes are ideal for light sensitive samples. Each box position is labeled with an alphanumeric grid for easy management of individual samples. 'No slide' features molded in bottom and lid provide secure stacking.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge