Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F188760020
BELAF188760020EA 75.1 EUR InStock
BELAF188760020
Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Gestelle Reagenzglasgestelle
This 20 place polypropylene round floating rack fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings.
  • Floats in water; can also be used for quickly thawing frozen cells in a 37 ºC water bath
  • Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
  • Dimensions: 6,4 mm (¹/₄") thick; 95 mm (3³/₄") in Ø; 19 mm (³/₄") long legs
  • Autoclavable
Excellent for holding cryotubes during work or incubation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge