Pipette and tip storage station, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F189619166
BELAF189619166EA 883 EUR InStock
BELAF189619166
Pipette and tip storage station, SP Bel-Art
Pipettenständer
This handy station keeps pipettors and tips within reach.
  • Top rack stores up to six pipettes; three bins hold a generous supply of bulk pipette tips
  • White acrylic plastic base, sides and top afford easy cleaning
  • Bin front and self-closing lid are made from transparent acrylic for clear view of contents
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge