Petri dish racks, Bel-Art ProCulture, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F189820109 F189820209 F189820309 F189820009 F189820314 F189820014 F189820409
BELAF189820109EA 510 EUR InStock
BELAF189820109 BELAF189820209 BELAF189820309 BELAF189820009 BELAF189820314 BELAF189820014 BELAF189820409
Petri dish racks, Bel-Art ProCulture, SP Bel-Art
Gestelle Spender für Petrischalen
Increase workspace and efficiently transport large quantities of Petri dishes.
  • Silicone holding strips grip each Petri dish; easily insert and remove groups of dishes without disturbing upper or lower dishes
  • Side labels provide space for critical sample identification
  • Lightweight, chemical resistant polycarbonate
  • Autoclavable at 121 ºC
Available in 4 colors and two sizes; the 241 mm high size holds forty-eight 100 mm Petri dishes, and the 368 mm high size holds eighty-four 100 mm Petri dishes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge