Vacuum aspiration systems, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F199170250
BELAF199170250EA 3320 EUR InStock
BELAF199170250
Vacuum aspiration systems, SP Bel-Art
Pumpen Vakuum-Absaugsysteme
These system is used to aspirate tissue culture media, supernatants, and chemical wastes.
  • Light weight, compact and easy to use; practical whenever a house vacuum is absent or not working
  • Hydrophobic filter protects pump from overflow
  • Low noise and minimal vibration
  • Chemical and corrosion resistant polypropylene collection bottle features heavy wall construction and a safety release vent to prevent vacuum implosion
  • Collection bottle, lid, tubing and connectors are all autoclavable at 121 °C; all held within a polypropylene pump housing
  • Hi-flow model has a stronger flow rate to work reliably with up to 8-channels, even with 4 channels empty
  • Hi-flow model includes quick disconnects to allow easy removal, emptying and replacement of the bottle
Built-in, maintenance free pump quickly and conveniently aspirates fluids.
Zertifizierungen: Hi-flow model ONLY (BELAF199170250) is CSA certified and the transformer includes an additional cord for 220 V type C outlets.
Lieferumfang: Complete system includes polypropylene bottle (3,78 L), tubing, safety vent and vaccum pump.
Achtung! Bitte beachten: Disposable pipette tip, pasteur pipette needs to be connect to collection tubing and plug into 110/120 V outlet.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge